4-Ethoxycarbonylphenylboronic acid, 97%
4-Ethoxycarbonylphenylboronic acid, 97%
4-Ethoxycarbonylphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Ethoxycarbonylphenylboronic acid, 97%

CAS: 4334-88-7 | C9H11BO4 | 193.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 367790250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4334-88-7
IUPAC Name[4-(ethoxycarbonyl)phenyl]boronic acid
Molecular FormulaC9H11BO4
InChI KeyZLNFACCFYUFTLD-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only