2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 97%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 97%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 97%
Thermo Scientific Chemicals

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 97%

CAS: 722-92-9 | C9H7F6NO | 259.15 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 367810250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS722-92-9
IUPAC Name2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC9H7F6NO
InChI KeyTZEJXCIGVMTMDY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F
さらに表示
Appearance (Color)Light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point147°C to 152°C
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only