N,N-Diisopropylethylamine, 99.5+%, Thermo Scientific Chemicals
N,N-Diisopropylethylamine, 99.5+%, Thermo Scientific Chemicals
N,N-Diisopropylethylamine, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diisopropylethylamine, 99.5+%, Thermo Scientific Chemicals

製品番号(カタログ番号) 367841000
価格(JPY)
-
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7087-68-5
IUPAC Nameethylbis(propan-2-yl)amine
Molecular FormulaC8H19N
InChI KeyJGFZNNIVVJXRND-UHFFFAOYSA-N
SMILESCCN(C(C)C)C(C)C
さらに表示
Appearance (Form)Clear liquid
GC>=99.50 %
Color scale=<50 APHA
Water=<0.3 %
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only