3-Carene, 90%, stabilized
3-Carene, 90%, stabilized
3-Carene, 90%, stabilized
Thermo Scientific Chemicals

3-Carene, 90%, stabilized

CAS: 13466-78-9 | C10H16 | 136.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
Packaging:
Glass bottle
Catalog number 367960250
Price (USD)
24.65
Online exclusive
27.70
Save 3.05
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
24.65
Online exclusive
27.70
Save 3.05
Each
Chemical Identifiers
CAS13466-78-9
IUPAC Name3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
Molecular FormulaC10H16
InChI KeyBQOFWKZOCNGFEC-UHFFFAOYNA-N
SMILESCC1=CCC2C(C1)C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Density(20°C) 0.862 to 0.865 g/ml
GC>=88 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only