Methyl linolenate, 99%
Methyl linolenate, 99%
Methyl linolenate, 99%
Thermo Scientific Chemicals

Methyl linolenate, 99%

CAS: 301-00-8 | C19H32O2 | 292.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 368100050
Price (USD)
334.65
Special offer
Online exclusive
Ends: 28-Dec-2026
393.00
Save 58.35 (15%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
334.65
Special offer
Online exclusive
Ends: 28-Dec-2026
393.00
Save 58.35 (15%)
Each
Chemical Identifiers
CAS301-00-8
IUPAC Namemethyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Molecular FormulaC19H32O2
InChI KeyDVWSXZIHSUZZKJ-YSTUJMKBSA-N
SMILESCC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=98.5 %
Refractive index1.4700 to 1.4720 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only