Glycerine trioleate, 98%
Glycerine trioleate, 98%
Glycerine trioleate, 98%
Thermo Scientific Chemicals

Glycerine trioleate, 98%

CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
25 mL
100 mL
Catalog number 368120010
Price (EUR)
47,65
Online exclusive
52,60
Save 4,95
Each
Quantity:
1 mL
Request bulk or custom format
Price (EUR)
47,65
Online exclusive
52,60
Save 4,95
Each
Chemical Identifiers
CAS122-32-7
IUPAC Name1,3-bis[(9Z)-octadec-9-enoyloxy]propan-2-yl (9Z)-octadec-9-enoate
Molecular FormulaC57H104O6
InChI KeyPHYFQTYBJUILEZ-IUPFWZBJSA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4680 to 1.4700 (20°C, 589 nm)
Appearance (Form)Liquid
HPLC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only