N-BOC-4-Hydroxypiperidine, 97%
N-BOC-4-Hydroxypiperidine, 97%
N-BOC-4-Hydroxypiperidine, 97%
Thermo Scientific Chemicals

N-BOC-4-Hydroxypiperidine, 97%

CAS: 109384-19-2 | C10H19NO3 | 201.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 368271000
Price (USD)
372.65
Special offer
Online exclusive
Ends: 28-Dec-2026
438.00
Save 65.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
372.65
Special offer
Online exclusive
Ends: 28-Dec-2026
438.00
Save 65.35
Each
Chemical Identifiers
CAS109384-19-2
IUPAC Nametert-butyl 4-hydroxypiperidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyPWQLFIKTGRINFF-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to cream
GC>=96.0 %
Melting point60°C to 65°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only