Methyl 3-(bromomethyl)benzoate, 95%
Methyl 3-(bromomethyl)benzoate, 95%
Methyl 3-(bromomethyl)benzoate, 95%
Thermo Scientific Chemicals

Methyl 3-(bromomethyl)benzoate, 95%

CAS: 1129-28-8 | C9H9BrO2 | 229.07 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 368631000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1129-28-8
IUPAC Namemethyl 3-(bromomethyl)benzoate
Molecular FormulaC9H9BrO2
InChI KeyYUHSMQQNPRLEEJ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
GC>=94 %
Melting point41°C to 47°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only