2-Chloro-4-picoline, 98%
2-Chloro-4-picoline, 98%
2-Chloro-4-picoline, 98%
Thermo Scientific Chemicals

2-Chloro-4-picoline, 98%

CAS: 3678-62-4 | C6H6ClN | 127.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 368840250
Price (TWD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3678-62-4
IUPAC Name2-chloro-4-methylpyridine
Molecular FormulaC6H6ClN
InChI KeyMZVSTDHRRYQFGI-UHFFFAOYSA-N
SMILESCC1=CC=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Refractive index1.5280 to 1.5300 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only