4-(Bromomethyl)biphenyl, 96%
4-(Bromomethyl)biphenyl, 96%
4-(Bromomethyl)biphenyl, 96%
Thermo Scientific Chemicals

4-(Bromomethyl)biphenyl, 96%

CAS: 2567-29-5 | C13H11Br | 247.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 368950050
Price (USD)
101.65
Special offer
Online exclusive
Ends: 28-Dec-2026
119.00
Save 17.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 28-Dec-2026
119.00
Save 17.35
Each
Chemical Identifiers
CAS2567-29-5
IUPAC Name4-(bromomethyl)-1,1'-biphenyl
Molecular FormulaC13H11Br
InChI KeyHZQLUIZFUXNFHK-UHFFFAOYSA-N
SMILESBrCC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point83°C to 86°C
Appearance (Color)Light yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only