(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, 97%
(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, 97%
(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, 97%
Thermo Scientific Chemicals

(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, 97%

CAS: 15186-48-8 | C6H10O3 | 130.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3692600101 gGlass bottle
Catalog number 369260010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15186-48-8
IUPAC Name(4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
Molecular FormulaC6H10O3
InChI KeyYSGPYVWACGYQDJ-YFKPBYRVSA-N
SMILESCC1(C)OC[C@@H](O1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoCrack to monomer by distillation before use
Appearance (Color)Clear to hazy colorless to yellow
Appearance (Form)Liquid or solid
GC>=96.0 %
Additional infoContains various amounts of polymer
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only