Chlorobis(cyclooctene)rhodium(I) dimer, 98%
Chlorobis(cyclooctene)rhodium(I) dimer, 98%
Chlorobis(cyclooctene)rhodium(I) dimer, 98%
Thermo Scientific Chemicals

Chlorobis(cyclooctene)rhodium(I) dimer, 98%

CAS: 12279-09-3 | C32H56Cl2Rh2 | 717.51 g/mol
数量:
100 mg
500 mg
パッケージ:
Glass bottle
製品番号(カタログ番号) 369375000
価格(JPY)
-
数量:
500 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12279-09-3
IUPAC Namebis(λ¹-rhodium(1+)) tetrakis((Z)-cyclooctene) dichloride
Molecular FormulaC32H56Cl2Rh2
InChI KeyGQPAPAIPOLEZHT-XFCUKONHSA-L
SMILES[Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1
さらに表示
Infrared spectrumConforms
Appearance (Color)Orange-brown
Appearance (Form)Powder
Rhodium (Rh)28.0 to 29.4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only