4-Acetylphenylboronic acid, 97%
4-Acetylphenylboronic acid, 97%
4-Acetylphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Acetylphenylboronic acid, 97%

CAS: 149104-90-5 | C8H9BO3 | 163.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 369400010
Price (USD)
28.30
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.30
Each
Chemical Identifiers
CAS149104-90-5
IUPAC Name(4-acetylphenyl)boronic acid
Molecular FormulaC8H9BO3
InChI KeyOBQRODBYVNIZJU-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only