Strontium chloride, 99.99%, (trace metal basis), anhydrous
Strontium chloride, 99.99%, (trace metal basis), anhydrous
Strontium chloride, 99.99%, (trace metal basis), anhydrous
Thermo Scientific Chemicals

Strontium chloride, 99.99%, (trace metal basis), anhydrous

CAS: 10476-85-4 | Cl2Sr | 158.53 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 369740250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10476-85-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
さらに表示
Appearance (Form)Crystalline powder
Total trace metal impurities=<0.01 %
Appearance (Color)White
Assay54.4 to 55.9 % (Sr)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only