PMSF Protease Inhibitor

Catalog number: 36978B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific PMSF is a protease inhibitor that reacts with serine residues to inhibit trypsin, chymotrypsin, thrombin and papain.

Features of PMSF:

• Targets serine proteases, including trypsin, chymotrypsin, thrombin and papain
• Effective in most protein solutions at 0.1 to 1 mM final concentration
• Because PMSF has limited water solubility, it must be dissolved in a small amount of solvent, such as ethanol, methanol or isopropyl alcohol before addition to a buffer

PMSF is an abbreviation for phenylmethylsulfonyl fluoride, the most common chemical name for this small compound. In addition to inhibiting serine proteases, PMSF will also inhibit cysteine proteases, like papain (reversible by DTT treatment), and mammalian acetylcholinesterase.

Properties of PMSF:
• Synonyms: alpha-Toluenesulfonyl fluoride; Benzylsulfonyl fluoride; Phenylmethylsulfonyl fluoride
• Formula: C6H5CH2SO2F
• Molecular weight: 174.19
• CAS Number: 329-98-6; EC Number: 206-350-2
• R/S Codes: R 25-34; S 26-36/37/39-45
• Storage conditions: room temperature in a dry place protected from light
• Working solution: dissolve PMSF in isopropanol at 1.74 mg/mL (10 mM); store in aliquots at -20°C.

Specifications of PMSF (Part No. 36978):
• Appearance: white powder
• Purity: > 99.0 %
• Melting point: 90-95°C
• Humidity: < 0.1 %

Related Products
Pierce™ Protease and Phosphatase Inhibitor Mini Tablets, EDTA Free
Pierce™ Protease Inhibitor Mini Tablets, EDTA Free
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Protease Inhibitor
Form: Liquid
Product Size: Custom

Contents & storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.

Documents

Manuals & protocols