2-Bromoterephthalic acid, 95%, Thermo Scientific Chemicals
2-Bromoterephthalic acid, 95%, Thermo Scientific Chemicals
2-Bromoterephthalic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoterephthalic acid, 95%, Thermo Scientific Chemicals

CAS: 586-35-6 | Molecular Formula: C8H5BrO4 | Molecular Weight: 245.03 g/mol
Have Questions?
Catalog NumberQuantity
36981025025 g
Catalog number 369810250
Price (USD)
203.65
Special offer
Online exclusive
Ends: 28-Dec-2026
239.00
Save 35.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
203.65
Special offer
Online exclusive
Ends: 28-Dec-2026
239.00
Save 35.35
Each
Chemical Identifiers
CAS586-35-6
IUPAC Name2-bromobenzene-1,4-dicarboxylic acid
Molecular FormulaC8H5BrO4
InChI KeyQPBGNSFASPVGTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Appearance (Color)White to light yellow
Appearance (Form)Powder
Titration with NaOH>=94 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only