Keratinozyten-SFM Supplement: Humaner rekombinanter epidermaler Wachstumsfaktor (EGF 1-53) & Rinder-Hypophysenextrakt (BPE)
Gibco™

Keratinozyten-SFM Supplement: Humaner rekombinanter epidermaler Wachstumsfaktor (EGF 1-53) & Rinder-Hypophysenextrakt (BPE)

Humaner rekombinanter epidermaler Wachstumsfaktor (EGF 1-53) und Rinderhypophysenextrakt (BPE).Weitere Informationen
Have Questions?
KatalognummerMenge
370000151 mL
Katalognummer 37000015
Preis (EUR)
108,00
Each
Menge:
1 mL
Preis (EUR)
108,00
Each
Humaner rekombinanter epidermaler Wachstumsfaktor (EGF 1-53) und Rinderhypophysenextrakt (BPE).

Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.

Specifications
ZelltypKeratinozyten, Epithelzellen
KulturtypSäugetierzellkultur
Menge1 mL
Serumspiegelserumfrei
Haltbarkeit18 Monate
VersandbedingungTrockeneis
SpeziesHuman
Klassifikationserumfrei
ProdukttypKeratinozyten-SFM Supplement
SterilitätSteril
Sterilization MethodSteril gefiltert
Mit AdditivenPhenol Red
Unit SizeEach
Inhalt und Lagerung
Im Gefrierschrank lagern (-5 bis -30 °C).

Häufig gestellte Fragen (FAQ)

Are the Keratinocyte SFM basal medium and supplement kit available separately?

The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which serum-free media can be used to culture mouse keratinocytes?

The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which serum-free media can be used to culture human keratinocytes?

There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

For how long can I store Keratinocyte-SFM at 4 degrees C?

The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.