Zinc acetate, 99%, pure
Zinc acetate, 99%, pure
Zinc acetate, 99%, pure
Thermo Scientific Chemicals

Zinc acetate, 99%, pure

CAS: 557-34-6 | C4H6O4Zn | 183.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 370080250
Price (USD)
112.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
112.00
Each
Chemical Identifiers
CAS557-34-6
IUPAC Namezinc(2+) diacetate
Molecular FormulaC4H6O4Zn
InChI KeyDJWUNCQRNNEAKC-UHFFFAOYSA-L
SMILES[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Infrared spectrumConforms
Titration Complexometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only