Sodium percarbonate, 13-14% active oxygen
Sodium percarbonate, 13-14% active oxygen
Sodium percarbonate, 13-14% active oxygen
Thermo Scientific Chemicals

Sodium percarbonate, 13-14% active oxygen

CAS: 15630-89-4 ∣ 1·5 H2O2 ∣ 157.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 370730025
Price (USD)
105.65
Special offer
Online exclusive
Ends: 28-Dec-2026
124.00
Save 18.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 28-Dec-2026
124.00
Save 18.35
Each
Chemical Identifiers
CAS15630-89-4
IUPAC Nametetrasodium tris(peroxol) dicarbonate
Molecular FormulaC2H6Na4O12
InChI KeyMSLRPWGRFCKNIZ-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].OO.OO.OO.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granular powder or granules
Appearance (Color)White
Titration with KMnO412.5 to 14.5 % (Available Oxygen)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

RUO – Research Use Only