Hematein
Hematein
Hematein
Thermo Scientific Chemicals

Hematein

CAS: 475-25-2 | C16H12O6 | 300.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 370840250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS475-25-2
IUPAC Name5,6,10,15-tetrahydroxy-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-1(17),2,4,6,12,15-hexaen-14-one
Molecular FormulaC16H12O6
InChI KeyHLUCICHZHWJHLL-UHFFFAOYNA-N
SMILESOC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Appearance (Form)Crystalline powder
Moisture=<15 %
Infrared spectrumConforms
UV spectrophotometry5 to 20 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only