Ethyl benzoylacetate, 90%
Ethyl benzoylacetate, 90%
Ethyl benzoylacetate, 90%
Thermo Scientific Chemicals

Ethyl benzoylacetate, 90%

CAS: 94-02-0 | C11H12O3 | 192.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 370855000
Price (USD)
239.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
239.00
Each
Chemical Identifiers
CAS94-02-0
IUPAC Nameethyl 3-oxo-3-phenylpropanoate
Molecular FormulaC11H12O3
InChI KeyGKKZMYDNDDMXSE-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5240 to 1.5330 (20°C, 589 nm)
HPLC>=88 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to red to brown
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only