Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Thermo Scientific Chemicals

Ammonium bicarbonate, 98%, pure

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 370930050
Price (USD)
135.00
Each
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
135.00
Each
Chemical Identifiers
CAS1066-33-7
IUPAC Namecarbonic acid amine
Molecular FormulaCH5NO3
InChI KeyATRRKUHOCOJYRX-UHFFFAOYSA-N
SMILESN.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HCl>=97.5 %
Appearance (Form)Crystalline powder
Anticaking agent=<0.55 % (MgCO3)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only