1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate
Thermo Scientific Chemicals

1-Bromo-2-(2-methoxyethoxy)ethane, 90%, stabilized with sodium carbonate

CAS: 54149-17-6 | C5H11BrO2 | 183.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 370980500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54149-17-6
IUPAC Name1-bromo-2-(2-methoxyethoxy)ethane
Molecular FormulaC5H11BrO2
InChI KeyHUXJXNSHCKHFIL-UHFFFAOYSA-N
SMILESCOCCOCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4540 to 1.4600 (20°C, 589 nm)
GC>=88 %
Infrared spectrumConforms
Stabilizer0.1 % Sodium carbonate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only