(R)-(+)-1-Phenylethylamine, 99+%
(R)-(+)-1-Phenylethylamine, 99+%
(R)-(+)-1-Phenylethylamine, 99+%
Thermo Scientific Chemicals

(R)-(+)-1-Phenylethylamine, 99+%

CAS: 3886-69-9 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 371460500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3886-69-9
IUPAC Name1-phenylethan-1-amine
Molecular FormulaC8H11N
InChI KeyRQEUFEKYXDPUSK-UHFFFAOYNA-N
SMILESCC(N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.5 %
Specific optical rotation+39° to +41° (20°C, 589 nm) (neat)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only