L-Asparagine, 99%
L-Asparagine, 99%
L-Asparagine, 99%
Thermo Scientific Chemicals

L-Asparagine, 99%

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number 371605000
Price (USD)
212.65
Special offer
Online exclusive
Ends: 28-Dec-2026
250.00
Save 37.35 (15%)
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
212.65
Special offer
Online exclusive
Ends: 28-Dec-2026
250.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS70-47-3
IUPAC Name2-amino-3-carbamoylpropanoic acid
Molecular FormulaC4H8N2O3
InChI KeyDCXYFEDJOCDNAF-UHFFFAOYNA-N
SMILESNC(CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 % (On dry substance)
Infrared spectrumConforms
Appearance (Color)White
Water=<1 %
Specific optical rotation+33° to +36° (20°C, 589 nm) (c=10, 2N HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only