Sodium hydroxide, pellets, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium hydroxide, pellets, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., ChP, U.S.P, B.P and E.P Specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
12 kg
Número de catálogo 3717-01
Precio (MXN)
-
Cantidad:
500 g

This product is GMP Manufactured and meets J.P., ChP, U.S.P, B.P and E.P Specifications.

Especificaciones
Punto de ebullición1390°C
CAS1310-73-2
Nombre del producto químico o materialSodium hydroxide
ColorWhite
Punto de fusión323°C
Fórmula molecularNaOH
EnvasePoly bottle
SinónimoSodium hydrate, Soda lye, Caustic soda, Lye
GradoMulti-Compendial/NF
Peso molecular (g/mol)40
Porcentaje de pureza95.0-101.0%
Cantidad500 g
Unit Size500 g