Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Methyl acetate, 99+%, Extra Dry, AcroSeal™

CAS: 79-20-9 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 371830010
Price (USD)
152.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
152.00
Each
Chemical Identifiers
CAS79-20-9
IUPAC Namemethyl acetate
Molecular FormulaC3H6O2
InChI KeyKXKVLQRXCPHEJC-UHFFFAOYSA-N
SMILESCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 %
GC>=99.0 %
Residue after evaporation=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only