SoftFit-L™ Non-Filtered Low Retention Pipette Tip in Reload Tower
SoftFit-L™ Non-Filtered Low Retention Pipette Tip in Reload Tower
Pipette samples with the accuracy and precision you expect from your pipette every time, with these low retention pipette tips that deliver maximum volume of sample.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호부피(미터법)멸균제품 사이즈
3721-05-RI20 μLSterileCase of 4800
3722-05-RI20 μLNon-sterileCase of 4800
3722-05-RIPK
3722-05-RI으로도 사용됨
20 μLNon-sterilePack of 960
3752-05-RI200 μLNon-sterileCase of 4800
3752-05-RIPK
RI3752-05-으로도 사용됨
200 μLNon-sterilePack of 960
3732-05-RI300 μLNon-sterileCase of 4800
3732-05-RIPK
3732-05-RI으로도 사용됨
300 μLNon-sterilePack of 960
3782-05-RI1000 μLNon-sterileCase of 3072
3782-05-RIPK
3782-05-RI으로도 사용됨
1000 μLNon-sterileEach
3721-05-RIPK
3721-05-RI으로도 사용됨
20 μLSterilePack of 960
3731-05-RI300 μLSterileCase of 4800
3731-05-RIPK
3731-05-RI으로도 사용됨
300 μLSterilePack of 960
3751-05-RI200 μLSterileCase of 4800
3751-05-RIPK
3751-05-RI으로도 사용됨
200 μLSterilePack of 960
카탈로그 번호 3721-05-RI
제품 가격(KRW)
556,000
온라인 행사
Ends: 31-Dec-2026
654,000
할인액 98,000
Case of 4800
부피(미터법):
20 μL
멸균:
Sterile
Unit Size:
Case of 4800
제품 가격(KRW)
556,000
온라인 행사
Ends: 31-Dec-2026
654,000
할인액 98,000
Case of 4800
Pipette samples with the accuracy and precision you expect from your pipette every time, with Thermo Scientific™ SoftFit-L™ pipette tips. Low retention tips deliver maximum volume of sample and are for use with Rainin™ LTS pipettes.
  • Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space
  • Compatible with SoftFit-L empty racks
  • Color-coded inserts allows quick identification of tip volume
사양
클린 클레임DNase, RNase, DNA, ATP, and Endotoxin Free
설명20L, Non-Filtered, Sterile, Refill
길이(영국식 단위)1.95 in.
길이(미터법)4.86cm
모델20L
색상Natural
색상 코드Red
필터링됨Non-filtered
용도 (장비)Rainin LTS LiteTouch
형식Refill
제품라인ART SoftFit L
멸균Sterile
팁 스타일Universal
부피(미터법)20 μL
Unit SizeCase of 4800