SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips

SoftFit-L™ Non-Filtered Low Retention Hinged Rack Pipette Tips

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityUnit Size
3722-05-HR20 μLNon-sterileCase of 4800
3721-05-HR20 μLSterileCase of 4800
3721-05-HRPK
also known as 3721-05-HR
20 μLSterilePack of 960
3751-05-HR200 μLSterileCase of 4800
3751-05-HRPK
also known as 3751-05-HR
200 μLSterilePack of 960
3752-05-HR200 μLNon-sterileCase of 4800
3731-05-HR300 μLSterileCase of 4800
3732-05-HR300 μLNon-sterileCase of 4800
3781-05-HR1000 μLSterileCase of 3072
3781-05-HRPK
also known as 3781-05-HR
1000 μLSterilePack of 768
3782-05-HR1000 μLNon-sterileCase of 3072
3782-05-HRPK
also known as 3782-05-HR
1000 μLNon-sterilePack of 768
Catalog number 3722-05-HR
Price (EUR)
-
Volume (Metric):
20 μL
Sterility:
Non-sterile
Unit Size:
Case of 4800
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Low-retention tips maximize sample delivery
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allows quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description20L, Non-Filtered, Non-sterile, Hinged Rack
Length (English)1.95 in.
Length (Metric)4.86cm
Model20L
AutoclavableAutoclavable
ColorNatural
Color CodeRed
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatHinged Rack
Product LineART SoftFit L
SterilityNon-sterile
Tip StyleLTS-style tip with positive stop
Volume (Metric)20 μL
Unit SizeCase of 4800