Matrix™ Open-Top Tri-coded Tubes

Matrix™ Open-Top Tri-coded Tubes

Green features
Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)멸균포장
3732-WP1D750 μLSterile10 Latch Racks of 96 Tubes/Cs.
3790-WP1D1400 μLNon-sterileBulk
3791-WP1D-BR1400 μLNon-sterile10 Latch Racks of 96 Tubes/Cs.
3792-WP1D-BR1400 μLSterile10 Latch Racks of 96 Tubes/Cs.
카탈로그 번호 3732-WP1D
제품 가격(KRW)
-
용량(미터법):
750 μL
멸균:
Sterile
포장:
10 Latch Racks of 96 Tubes/Cs.
Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side. Tubes are available in bulk packs or in barcoded latch racks. Barcoded latch racks feature a linear barcode, 2D barcode, and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific™ Matrix™ SepraSeal and DurSeal capping systems and the Thermo Scientific™ VisionMate™family of barcode readers.

Secure Tracking

  • Each polypropylene open-top tube has a unique 2D barcode laser-etched onto the base, plus a white patch with 1D linear barcode and human readable text on the side
  • 2D barcodes are resistant to 100% DMSO, ideal for compound processing and storage
  • Non-proprietary 2D barcodes and 1D linear barcodes can be scanned with the Thermo Scientific VisionMate family of barcode readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Barcoded latch racks include 1D and 2D barcodes, plus human readable text on three sides to provide added same traceability
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to guarantee readability
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes

Storage Tracking Solutions for Any Laboratory

  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
  • Tubes are available with a range of sealing options including Thermo Scientific SepraSeals and DuraSeals
  • SepraSeals are available in a range of color options for visual categorization of samples
  • Pre-Split SepraSeals allow for repeated sample access in automation

Recommended for:

Drug discovery, compound storage, secure sample transport, individual sample retrieval, instant sample identification, secure tracking of archived samples

사양
바코드Tube barcode: Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction and Linear EAN Code 128
색상Colorless
설명2D Barcoded Open-Top Tubes with White Patch and Linear Barcode in Latch Racks
그린 기능ACT Ecolabel (Gold)
라벨 유형2D White Patch
포장10 Latch Racks of 96 Tubes/Cs.
포장 수량10 Latch Racks of 96 Tubes/Cs.
제품라인Matrix
오토클레이브 가능Not Autoclavable
하단 형상Conical
용량(미터법)750 μL
GraduatedNo
물질Polypropylene
제품 유형Storage Tube (Open Top)
멸균Sterile
Unit SizeCase of 960