Dense Storage Options
Dense Storage Options
Dense Storage Options
Dense Storage Options
Thermo Scientific™
Dense Storage Options
Double your cryobank capacity and cut storage costs by 50%. Our dense storage options fit standard cryogenic storage boxes that work with the freezer racks you currently own.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
374001Cryobox Divider
5050-0001Undivided CryoBox
5050-0169Cryobox
374115Cryobank Tubes
Catalog number 374001
Price (BRL)
-
Type:
Cryobox Divider
Double your cryobank storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard CryoBoxes™ with 13 x 13 tube separators and fit 169 1mL tubes in a single CryoBox™. These standard-sized boxes are compatible with existing standard-sized lab freezer racks.
  • Choose a standard-sized cardboard or polycarbonate box
  • Insert the 13x13 box divider
  • Load the box with 169, 1mL Cryobank storage tubes
  • Slide the box into your existing standard-sized CryoBox racks
  • Suitable for mechanical freezer or vapor phase liquid nitrogen storage
Specifications
DescriptionCryobox divider, 13 x 13 grid
Quantity24/Cs.
Capacity (Metric)0.5 mL (Tube), 1 mL (Tube)
ColorBlack, White
Holds169 1 mL Nunc Cryobank Tubes
MaterialPolycarbonate
TypeCryobox Divider
Vial TypeCryobank
Unit SizeCase of 24