Matrix™ ScrewTop Tri-coded Tubes
Thermo Scientific™

Matrix™ ScrewTop Tri-coded Tubes

Green features
Securely store precious samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μl and 1.0mL ScrewTop Tubes.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)EsterilidadTipo de producto
3741-WP1D< 1,0/mlEstérilTapa de rosca con etiqueta blanca
3743-WP1D0,5 mlEstérilTubo con tapón de rosca
3744-WP1D-BR0,5 mlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3744-WP1D500 μlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3745-WP1D-BR0,5 mlEstérilTubo con tapón de rosca, Código de barras 2D
3745-WP1D500 μlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3740-WP1D< 1,0/mlEstérilTubo con tapón de rosca
3741-WP1D-BR< 1,0/mlEstérilTapa de rosca con etiqueta blanca
3742-WP1D< 1,0/mlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3742-WP1D-BR< 1,0/mlEstérilTubo con tapón de rosca, Código de barras lineal
3741NS-WP1D-BR1,1 mLNo estérilTapa de rosca con etiqueta blanca
3744NS-WP1D-BR0,5 mLNo estérilTapa de rosca con etiqueta blanca
3745NS-WP1D-BR0,5 mLNo estérilTapa de rosca con etiqueta blanca
Número de catálogo 3741-WP1D
Precio (CLP)
-
Capacidad (métrico):
< 1,0/ml
Esterilidad:
Estéril
Tipo de producto:
Tapa de rosca con etiqueta blanca
Securely store precious samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 0.64 mL  and 1.1 mL ScrewTop Tubes. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D,1D linear and human readable barcode. These tubes can be ordered to come in racks (barcoded or non-barcoded) or may be purchased in bulk. The tri-coded tubes can be purchased sterile or non-sterile. We also offer tubes with white patch only if your work does not require the 1D linear and human readable code. The 2D screwtop tubes meet a range of manual and automated long term storage needs; barcoded racks make it easy to track the rack as well. Use in combination with Thermo Scientific barcode readers, arrayers and capping equipment.

Features:

Dependable Sample Protection

  • Tubes are constructed of virgin class VI medical grade polypropylene
  • Screw-top caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • Screw-top tubes are compatible with storage down to -176°C

Secure Tracking

  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping/decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time

Stringent Quality Control

  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All sterile tubes are supplied free from DNA, RNase, DNase, PCR Inhibitors, endotoxins and cytotoxins
  • All non-sterile tubes are supplied free of RNase, DNase and endotoxins
Especificaciones
Código de barrasMatriz de datos 12 x 12 no exclusiva con corrección de errores integrada ECC200
Capacidad (imperial)0,0338 fl oz
Color del tapónIncolora
Material del cierrePolipropileno puro de calidad médica clase VI con junta de elastómero termoplástico moldeado
Sin ADNasaSí
DescripciónTubos con cuello de rosca y fondo en V con etiqueta blanca y código de barras lineal
Características ecológicasACT Ecolabel (Gold)
Envase5 Latch Racks of 96 Tubes
Línea de productosMatrix
Forma del pocilloFondo en V
Esterilizables en autoclaveEsterilizables en autoclave (con tapón suelto)
Forma del fondoCónicos
Capacidad (métrico)< 1,0/ml
Fit for PurposeSterile and PCR Ready
MaterialPolipropileno virgen de calidad médica Clase VI
Tipo de productoTapa de rosca con etiqueta blanca
EsterilidadEstéril
Unit SizeCase of 480