Matrix™ 1.1 mL V-Bottom ScrewTop Tubes, white patch/2D barcode, Clear/Colorless - FAQs
Matrix™ 1.1 mL V-Bottom ScrewTop Tubes, white patch/2D barcode, Clear/Colorless - FAQs
View additional product information for Matrix™ ScrewTop Tri-coded Tubes - FAQs (3741NS-WP1D-BR, 3743-WP1D, 3744-WP1D-BR, 3740-WP1D, 3745-WP1D-BR, 3742-WP1D, 3745-WP1D, 3744-WP1D, 3742NS-WP1D, 3742-WP1D-BR, 3745NS-WP1D-BR, 3744NS-WP1D-BR, 3741-WP1D-BR, 3741-WP1D)
2 product FAQs found
How do you prevent 2D barcode duplication during manufacturing and ensure quality control of the Thermo Scientific 2D barcoded storage tubes?
The 2D barcode laser etching software includes algorithms that prevent the barcode generator from ever counting backwards, subsequently preventing the re-etching of any previously generated barcode. To verify the above procedure, several quality control checks are performed on the 2D barcodes including:
- Checking for light transmission through the barcode as light transmission will reduce readability
- Inspecting the barcode under magnification
- Reading the barcode on a full rack 2D barcode reader
- Checking the barcode against a database of previously etched 2D barcodes
As the above quality control steps are completed, defective storage tubes are discarded, replaced, and re-checked, ensuring that the 2D barcodes are 100% readable and have not been previously used. This quality control process is the reason why we cannot guarantee sequential 2D barcode printing within a single rack of 2D barcoded products.
Can the barcodes on the Thermo Scientific 2D barcoded storage tubes be damaged by chemicals or by exposure to extreme temperatures?
The Thermo Scientific 2D barcoded storage tubes have been subjected in-house to consecutive liquid nitrogen freeze-thaws followed by submersion in boiling water. These tests did not cause any damage to the 2D barcode.
The 2D barcodes are made of the same material as the storage tubes and are resistant to standard lab chemicals.
The 2D barcode has a built-in ECC200 error barcode correction, which ensures that a damaged barcode is still readable as long as less than 25% of the barcode is missing.