Nunc™ Low Profile 5 mL Externally-Threaded Universal Tubes, 2D barcode
Nunc™ Low Profile 5 mL Externally-Threaded Universal Tubes, 2D barcode
Nunc™ Low Profile 5 mL Externally-Threaded Universal Tubes, 2D barcode
Nunc™ Low Profile 5 mL Externally-Threaded Universal Tubes, 2D barcode
Thermo Scientific™
Nunc™ Low Profile 5 mL Externally-Threaded Universal Tubes, 2D barcode
Meet your tracking and packaging requirements for solid or liquid sample storage with these low-profile, universal tubes. Available in clear and color-coded caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorBarcodePackaging
374320RED-BRRed2D, Barcoded Rack10 Racks of 24 Tubes
374320REDRed2D10 Racks of 24 Tubes
374320ORGOrange2D10 Racks of 24 Tubes
374320YELYellow2D10 Racks of 24 Tubes
374320GRNGreen2D10 Racks of 24 Tubes
374320BLUBlue2D10 Racks of 24 Tubes
374320PURPurple2D10 Racks of 24 Tubes
374320ORG-BROrange2D, Barcoded Rack10 Racks of 24 Tubes
374320YEL-BRYellow2D, Barcoded Rack10 Racks of 24 Tubes
374320GRN-BRGreen2D, Barcoded Rack10 Racks of 24 Tubes
374320BLU-BRBlue2D, Barcoded Rack10 Racks of 24 Tubes
374320PUR-BRPurple2D, Barcoded Rack10 Racks of 24 Tubes
374322REDRed2D10 Bags of 24 Tubes
374322ORGOrange2D10 Bags of 24 Tubes
374322YELYellow2D10 Bags of 24 Tubes
374322GRNGreen2D10 Bags of 24 Tubes
374322BLUBlue2D10 Bags of 24 Tubes
374322PURPurple2D10 Bags of 24 Tubes
Catalog number 374320RED-BR
Price (EUR)
-
Closure Color:
Red
Barcode:
2D, Barcoded Rack
Packaging:
10 Racks of 24 Tubes
Meet your tracking and packaging requirements for solid or liquid sample storage with Thermo Scientific™ Nunc™ Low-Profile 5 mL Externally-Threaded Universal Tubes. A robust solution for solid or liquid sample storage, tubes feature 2D barcodes, graduations, a white patch for labeling, and a wide-mouth design for easy addition and extraction of sample material. The externally threaded design minimizes contamination risk in manual handling procedures. The tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures.
Flexible Tube Design
  • Wide mouth provides easy access using forceps for adding approximately (3) 1 mm sections of tissue, plant material or other solid sample for storage
  • Overall tube height of less than 50 mm and U-bottom shape allows convenient access with standard length pipette tips when used for liquid storage
  • Available with or without a 2D barcode
  • Include printed graduations and white writable surface for manual sample identification
  • Can be purchased in universal style latch rack or in bulk to meet your sample handling requirements
Cap Design
  • The cap features automation-friendly screw top that is compatible with Nunc capping and decapping equipment
  • The tube with the externally threaded cap is designed for cryostorage down to the vapor phase of liquid nitrogen
Secure Tracking
  • Permanently bonded, unique 2D barcode is laser-etched on base of tube to securely identify and track samples
  • Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes
  • Thermo Scientific 2D barcode reader instantly decodes each tube's barcode, transferring tube information into your application or database
  • Tubes include white writable space for sample information in addition to, or instead of, using 2D barcodes
Stringent Quality Control
  • 2D barcodes are scanned after manufacturing to ensure readability
  • Barcodes are checked against database of previously assigned codes to prevent duplicates
  • Tubes are leak-tested to ensure sample integrity and security
Specifications
Barcode2D, Barcoded Rack
Closure ColorRed
Packaging10 Racks of 24 Tubes
AutoclavableNot Autoclavable
Bottom ShapeFlat
GraduatedYes
MaterialPolypropylene
Product TypeCryogenic Storage Tube
SterilitySterile
Thread StyleExternal
Unit SizeCase of 10