Matrix™ ScrewTop Tri-coded Tubes
Thermo Scientific™

Matrix™ ScrewTop Tri-coded Tubes

Green features
Securely store precious samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μl and 1.0mL ScrewTop Tubes.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)EsterilidadTipo de producto
3745-WP1D500 μlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3744-WP1D-BR0,5 mlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3744-WP1D500 μlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3745-WP1D-BR0,5 mlEstérilTubo con tapón de rosca, Código de barras 2D
3741-WP1D-BR< 1,0/mlEstérilTapa de rosca con etiqueta blanca
3741-WP1D< 1,0/mlEstérilTapa de rosca con etiqueta blanca
3742-WP1D< 1,0/mlEstérilTapa de rosca con etiqueta blanca, Código de barras lineal
3742-WP1D-BR< 1,0/mlEstérilTubo con tapón de rosca, Código de barras lineal
3741NS-WP1D-BR1,1 mLNo estérilTapa de rosca con etiqueta blanca
3744NS-WP1D-BR0,5 mLNo estérilTapa de rosca con etiqueta blanca
3745NS-WP1D-BR0,5 mLNo estérilTapa de rosca con etiqueta blanca
Número de catálogo 3745-WP1D
Precio (USD)
-
Capacidad (métrico):
500 μl
Esterilidad:
Estéril
Tipo de producto:
Tapa de rosca con etiqueta blanca, Código de barras lineal
Securely store precious samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 0.64 mL  and 1.1 mL ScrewTop Tubes. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D,1D linear and human readable barcode. These tubes can be ordered to come in racks (barcoded or non-barcoded) or may be purchased in bulk. The tri-coded tubes can be purchased sterile or non-sterile. We also offer tubes with white patch only if your work does not require the 1D linear and human readable code. The 2D screwtop tubes meet a range of manual and automated long term storage needs; barcoded racks make it easy to track the rack as well. Use in combination with Thermo Scientific barcode readers, arrayers and capping equipment.

Features:

Dependable Sample Protection

  • Tubes are constructed of virgin class VI medical grade polypropylene
  • Screw-top caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • Screw-top tubes are compatible with storage down to -176°C

Secure Tracking

  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping/decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time

Stringent Quality Control

  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All sterile tubes are supplied free from DNA, RNase, DNase, PCR Inhibitors, endotoxins and cytotoxins
  • All non-sterile tubes are supplied free of RNase, DNase and endotoxins
Especificaciones
Código de barrasMatriz de datos 12 x 12 no exclusiva con corrección de errores integrada ECC200
Capacidad (imperial)16907 fl oz
Color del tapónn.a.
Cierre incluidoNo
Sin ADNasaSí
DescripciónTubos con cuello de rosca y fondo en V con etiqueta blanca y código de barras lineal
Características ecológicasACT Ecolabel (Gold)
EnvaseCaja
Línea de productosMatrix
Forma del pocilloFondo en V
Forma del fondoCónicos
Capacidad (métrico)500 μl
Fit for PurposeSterile and PCR Ready
MaterialPolipropileno virgen de calidad médica Clase VI
Tipo de productoTapa de rosca con etiqueta blanca, Código de barras lineal
EsterilidadEstéril
Unit SizeCase of 480

Preguntas frecuentes

What are the main advantages of the Thermo Scientific 2D barcoded storage system?

Here are some advantages of the 2D barcoded storage system:
- Less time needed for labeling, sample recording, and retrieval
- Maximize freezer utilization while keeping storage space organised
- Ensures a safe, permanent ID of a sample
- Enables multivial capping and multichannel filling
- Individualized samples, eliminating multiple freeze/thaw cycles

Why do you recommend Nunc Cryobank vials for liquid nitrogen storage and the Matrix ScrewTop storage tubes for storage at higher temperatures?

We recommend using Nunc Cryobank vials for liquid nitrogen storage as these vials are designed for use at ultra-low temperatures. They come without a lid so that they can be easily picked from the rack at ultra-low temperatures using a tube selection or pen tool.
On the other hand, we recommend Matrix ScrewTop storage tubes for use at higher temperatures (-80 degrees C and above). They work well for both manual and automated picking.

Which Thermo Scientific 2D barcoded storage tubes should I use for my application?

Please view the following information regarding the usage of Thermo Scientific 2D barcoded storage tubes:
Tube Type; Typical Storage Temperature; Application(s); Special Feature(s)
Sepraseal; -20 degrees C; Compound storage; Can easily be removed for sample access
Presplit Sepraseal; -20 degrees C; Compound storage; Up to 200 piercings
Duraseal; -20 degrees; Compound storage; Safe seal for multiple piercings
Matrix ScrewTop; -80 degrees C; Blood components, DNA, Bacteria; With the correct torque, there is very little evaporation at -80 C, disposable pipette tips can be used
Nunc Screwcap; -176 degrees C; Mammalian cells; Minimal height, tubes can be removed with cryo gloves