Nunc™ 1.8mL Externally-Threaded Universal Tubes, non-coded, in 1D/2D barcoded latch rack - FAQs
View additional product information for Nunc™ 1.8mL Externally-Threaded Universal Tubes and Racks - FAQs (374501, 374502, 374500TS, 374501-BR, 374500-BR, 374502NOV, 374503)
1 product FAQs found
What is the difference between Nunc 1.8mL Externally-Threaded Universal Tubes and Racks (Cat. Nos. 374500-BR and 374501-BR)?
Catalog number 374500-BR is our Nunc universal tubes with 2D code in a barcoded latch rack.
Catalog number 374501-BR is our Nunc universal tubes without 2D code in a barcoded latch rack.