Nunc™ 1.8 mL Externally-Threaded, Sterile Universal Tubes, Bulk pack
Thermo Scientific™
Nunc™ 1.8 mL Externally-Threaded, Sterile Universal Tubes, Bulk pack
Use the automation friendly tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available in clear and color-coded caps, racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColor del tapónCódigo de barrasEnvase
374502ORGOrange2D10 Bags of 48 Tubes
374502REDRed2D10 Bags of 48 Tubes
374502YELYellow2D10 Bags of 48 Tubes
374502GRNGreen2D10 Bags of 48 Tubes
374502BLUBlue2D10 Bags of 48 Tubes
374502PURPurple2D10 Bags of 48 Tubes
374503REDRedNon-coded10 Bags of 48 Tubes
374503ORGOrangeNon-coded10 Bags of 48 Tubes
374503YELYellowNon-coded10 Bags of 48 Tubes
374503GRNGreenNon-coded10 Bags of 48 Tubes
374503BLUBlueNon-coded10 Bags of 48 Tubes
374503PURPurpleNon-coded10 Racks of 48 Tubes
Número de catálogo 374502ORG
Precio (MXN)
-
Color del tapón:
Orange
Código de barras:
2D
Envase:
10 Bags of 48 Tubes
Use the automation friendly Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes to improve the efficiency and throughput of your sample storage workflow. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.
Scalable
  • Tubes are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10 x 10 arrays
  • External threads limit contamination
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
  • 374502NOV is featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
Especificaciones
Código de barras2D
Color del tapónOrange
Envase10 Bags of 48 Tubes
Esterilizables en autoclaveNot Autoclavable
Forma del fondoFlat
GraduatedYes
MaterialPolypropylene
Tipo de productoCryogenic Storage Tube
EsterilidadSterile
Estilo de roscaExternal
Unit SizeCase of 480