Nunc™ 2 mL Internally-Threaded Universal Tubes with Universal latch rack, 2D code
Thermo Scientific™
Nunc™ 2 mL Internally-Threaded Universal Tubes with Universal latch rack, 2D code
Experience the best features of our storage tube portfolio: universal cap, universal latch rack and free-standing tube design -- all for either benchtop or automation use. Available in clear and color-coded caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorBarcodePackaging
374516GRNGreen2D10 Bags of 48 Tubes
374510REDRed2D10 Racks of 48 Tubes
374510ORGOrange2D10 Racks of 48 Tubes
374510YELYellow2D10 Racks of 48 Tubes
374510GRNGreen2D10 Racks of 48 Tubes
374510BLUBlue2D10 Racks of 48 Tubes
374510PURPurple2D10 Racks of 48 Tubes
374510RED-BRRed2D, Barcoded Rack10 Racks of 48 Tubes
374510ORG-BROrange2D, Barcoded Rack10 Racks of 48 Tubes
374510YEL-BRYellow2D, Barcoded Rack10 Racks of 48 Tubes
374510GRN-BRGreen2D, Barcoded Rack10 Racks of 48 Tubes
374510BLU-BRBlue2D, Barcoded Rack10 Racks of 48 Tubes
374510PUR-BRPurple2D, Barcoded Rack10 Racks of 48 Tubes
374511REDRedNon-coded10 Racks of 48 Tubes
374511ORGOrangeNon-coded10 Racks of 48 Tubes
374511YELYellowNon-coded10 Racks of 48 Tubes
374511GRNGreenNon-coded10 Racks of 48 Tubes
374511BLUBlueNon-coded10 Racks of 48 Tubes
374511PURPurpleNon-coded10 Racks of 48 Tubes
374511RED-BRRedBarcoded Rack10 Racks of 48 Tubes
374511ORG-BROrangeBarcoded Rack10 Racks of 48 Tubes
374511YEL-BRYellowBarcoded Rack10 Racks of 48 Tubes
374511GRN-BRGreenBarcoded Rack10 Racks of 48 Tubes
374511BLU-BRBlueBarcoded Rack10 Racks of 48 Tubes
374511PUR-BRPurpleBarcoded Rack10 Racks of 48 Tubes
374516REDRed2D10 Bags of 48 Tubes
374516ORGOrange2D10 Bags of 48 Tubes
374516YELYellow2D10 Bags of 48 Tubes
374516BLUBlue2D10 Bags of 48 Tubes
374516PURPurple2D10 Bags of 48 Tubes
Catalog number 374516GRN
Price (USD)
-
Closure Color:
Green
Barcode:
2D
Packaging:
10 Bags of 48 Tubes
Experience the best features of the Thermo Scientific™ storage tube portfolio: universal cap, universal latch rack and free-standing tube. This versatile, 2 mL internally threaded tube is ideal both for a large storage system like a biobank, and in smaller scale for traditional cryostorage. Sample identity is ensured due to a permanently attached 2D barcode on each tube and permanent laser etching of the rack. Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes.
Scalable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
Specifications
Barcode2D
Closure ColorGreen
Packaging10 Bags of 48 Tubes
AutoclavableNot Autoclavable
Bottom ShapeFlat
GraduatedYes
MaterialPolypropylene
Product TypeCryogenic Storage Tube
SterilitySterile
Thread StyleInternal
Unit SizeCase of 480