2-Iodoxybenzoic acid, stabilized
2-Iodoxybenzoic acid, stabilized
2-Iodoxybenzoic acid, stabilized
Thermo Scientific Chemicals

2-Iodoxybenzoic acid, stabilized

CAS: 61717-82-6 | C7H5IO4 | 280.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 374650500
Price (USD)
588.65
Special offer
Online exclusive
Ends: 28-Dec-2026
692.00
Save 103.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
588.65
Special offer
Online exclusive
Ends: 28-Dec-2026
692.00
Save 103.35
Each
Chemical Identifiers
CAS61717-82-6
IUPAC Name1-hydroxy-1-oxo-3H-1λ⁵,2-benziodaoxol-3-one
Molecular FormulaC7H5IO4
InChI KeyCQMJEZQEVXQEJB-UHFFFAOYSA-N
SMILESO[I]1(=O)OC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder or crystals
IdentificationIsophthalic acid, Benzoic acid)
Titration Iodometric39 to 50 % as 2-Iodoxybenzoic acid
IdentificationHPLC (2-Iodoxybenzoic acid, 2-Iodosobenzoic acid,
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
RUO – Research Use Only