Dimethyl aminomalonate hydrochloride, 97%
Dimethyl aminomalonate hydrochloride, 97%
Dimethyl aminomalonate hydrochloride, 97%
Thermo Scientific Chemicals

Dimethyl aminomalonate hydrochloride, 97%

CAS: 16115-80-3 | C5H10ClNO4 | 183.59 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 374970250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16115-80-3
IUPAC Namehydrogen 1,3-dimethyl 2-aminopropanedioate chloride
Molecular FormulaC5H10ClNO4
InChI KeyQWNDKNJSEWOEDM-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC(=O)C(N)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Color)Light yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only