Triethyl citrate, 99%
Triethyl citrate, 99%
Triethyl citrate, 99%
Thermo Scientific Chemicals

Triethyl citrate, 99%

CAS: 77-93-0 | C12H20O7 | 276.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 375060025
Price (USD)
234.65
Special offer
Online exclusive
Ends: 28-Dec-2026
276.00
Save 41.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
234.65
Special offer
Online exclusive
Ends: 28-Dec-2026
276.00
Save 41.35
Each
Chemical Identifiers
CAS77-93-0
IUPAC Name1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular FormulaC12H20O7
InChI KeyDOOTYTYQINUNNV-UHFFFAOYSA-N
SMILESCCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Clear liquid
Refractive index1.4400 to 1.4430 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only