SuperBlock™ (PBS) Blocking Buffer

Catalog number: 37515B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific SuperBlock (PBS) Blocking Buffer is optimized PBS solution containing a protein that provides incredibly fast and effective blocking for ELISA, IHC and Western blot analysis.

Features of SuperBlock (PBS) Blocking Buffer:

Fast—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
Flexible—guaranteed to be biotin-free for use with streptavidin system
Convenience—available in multiple package sizes (100 mL, 1L and 5L)
Low background—the non-serum protein solution yields a high signal-to-noise ratio
Stable—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

Related Products
SuperBlock™ T20 (PBS) Blocking Buffer
SuperBlock™ (TBS) Blocking Buffer
SuperBlock™ T20 (TBS) Blocking Buffer
SuperBlock™ (TBS) Blocking Buffer Dry Blend
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: SuperBlock™
Product Size: Custom
Reagent Type: Blocking Reagent
Validated Application: ELISA, Western Blot

Contents & storage

Upon receipt store at 4°C. Product is shipped at ambient temperature.

Documents

Manuals & protocols