This product is only available through your local Thermo Scientific supplier. Learn where to buy >
SEA BLOCK Blocking Buffer
Catalog number: 37527B
Thermo Scientific™  Related applications: Protein Assays and Analysis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific SEA BLOCK Blocking Buffer is steelhead salmon serum in PBS that is especially useful as a blocking agent in IHC and other detection methods involving mammalian samples.
Features of SEA BLOCK Blocking Buffer:
Non-mammalian—fish proteins are less likely to have specific binding interactions with antibodies and other mammalian proteins present in typical methods
Convenient—filtered and stabilized in PBS for compatibility with most assay systems
Easy to use—can be used as supplied or diluted up to 10-fold as needed
Flexible—may be used for many different applications, including as a diluent for antibodies
Because SEA BLOCK Blocking Buffer is fish serum, it is effective in stabilizing solutions and samples for antibody-binding interactions while at the same time minimizing the possibility of cross-reaction with mammalian sample components. When normal sera cause background in immunohistochemistry (IHC), try this product. The product is also especially effective in various cellular imaging protocols based on visible and near-infrared fluorescence detection.
Fish serum has several advantages over typical blocking buffers. Because salmon is phylogenetically distant from mammals, which are the source of antibodies and samples used in most experiments, its serum proteins are less likely to have specific binding interactions with proteins used in typical cell biology experiments. Because SEA BLOCK Blocking Buffer is fish serum, it is effective in stabilizing solutions and samples for antibody-binding interactions. When normal sera cause background in IHC experiments, SEA BLOCK Blocking Buffer may correct the problem.
SEA BLOCK Blocking Buffer is particularly effective in applications involving fluorescence imaging. The blocker has been used to decrease background and increase signal-to-noise ratios with near-IR fluorescent probes and is validated for use with the LI-COR Odyssey™ Infrared Imaging System.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Form: Liquid
Product Size: Custom
Reagent Type: Blocking Reagent
Validated Application: ELISA, Western Blot
Contents & storage
Upon receipt store at 4°C.
Documents
Manuals & protocols