Blocker™ Casein in TBS

Catalog number: 37532B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Blocker Casein in TBS is a ready-to-use, TBS solution of purified casein protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.

This blocking buffer contains casein protein that is purified from milk by the Hammarsten method. The phosphate- and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody-based detection methods. Blocker Casein Buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features:

Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solutions are ready to use; can be diluted further as needed
Flexible—available in PBS and TBS formulations to suit a variety of applications
Safe—stable, thimerosal-free formulations

Related Products
Blocker™ Casein in PBS
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Blocker™
Product Size: Custom
Reagent Type: Blocking Reagent
Validated Application: ELISA, Western Blot

Contents & storage

Store at 4°C.

Documents

Manuals & protocols