Guardian™ Peroxidase Conjugate Stabilizer/Diluent
Guardian™ Peroxidase Conjugate Stabilizer/Diluent
Guardian™ Peroxidase Conjugate Stabilizer/Diluent
Guardian™ Peroxidase Conjugate Stabilizer/Diluent
Thermo Scientific™

Guardian™ Peroxidase Conjugate Stabilizer/Diluent

Thermo Scientific Guardian Peroxidase Conjugate Stabilizer/Diluent preserves the functional integrity and activity of horseradish peroxidase conjugated antibodies and other proteins자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
37548200 mL
375521 L
카탈로그 번호 37548
제품 가격(KRW)
205,000
온라인 행사
Ends: 30-Jun-2026
241,000
할인액 36,000 (15%)
Each
수량:
200 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
205,000
온라인 행사
Ends: 30-Jun-2026
241,000
할인액 36,000 (15%)
Each
Thermo Scientific Guardian Peroxidase Conjugate Stabilizer/Diluent preserves the functional integrity and activity of horseradish peroxidase conjugated antibodies and other proteins at very dilute concentrations for long-term storage.

Features of Guardian Peroxidase Conjugate Stabilizer/Diluent:

Preserves HRP activity—no significant loss of HRP activity over a six-month period at room temperature (1:1000 dilution) or 12 months at 4°C
Convenient—store ready-to-use dilutions (1:1000 to 1:100,000) that maintain enzyme activity in the refrigerator—no aliquoting or freezing
Assay compatible—simply add your favorite blocking buffer to create the ideal diluent for your HRP-based ELISA system or store the HRP conjugate as a 1:1000 stock solution for Western blots and dilute in the final assay buffer
Saves money—less expensive than competitor formulations or ordering new HRP conjugates

With Guardian Peroxidase Conjugate Stabilizer/Diluent, typical 1 mg/mL antibody or streptavidin peroxidase conjugates can be diluted as much as 100,000-fold for storage at 4°C. Most ELISA and blotting applications for HRP conjugates require dilution of typical 1 mg/mL stocks by at least 1000-fold (to 1 μg/mL). Sensitive assay systems, such as with chemiluminescent substrates, often require dilution up to 100,000-fold (10 ng/mL). The Guardian Solution enables these 1X working concentrations of HRP conjugate to be prepared in advanced and stored at 4°C for 12 months or at room temperature for six months.For blotting applications, it is usually best to prepare a concentration for storage that can be diluted 10-fold in assay buffer (which may contain blocking components) for final use.

Typical colorimetric ELISA procedures requires that 10 mL of HRP-conjugated secondary antibody be diluted 1000- to 5000-fold from a 1 mg/mL stock solution. That necessitates pipetting 2 to 10 μL of stock into a tube with 10 mL of diluent. More sensitive assay systems, such as Western blots with Thermo Scientific SuperSignal West Pico, Dura or Femto Chemiluminescent Substrates for HRP, require a second round of dilution to achieve the needed working concentration of HRP conjugate. With Guardian Stabilizer-Diluent, HRP conjugates can be pre-diluted and stored at concentrations that are either ready to use or easily diluted 10- to 100-fold for final use. Preparing larger volumes of pre-diluted stocks ensures consistency and greater accuracy for assays performed repeatedly day after day.
For Research Use Only. Not for use in diagnostic procedures.
사양
화학물질 이름 또는 재질Peroxidase Conjugate Stabilizer/Diluent
권장 스토리지Upon receipt store at room temperature or 4°C.
물리적 형태Liquid
제품라인Guardian
수량200 mL
Unit SizeEach