Guardian™ Peroxidase Conjugate Stabilizer/Diluent

Catalog number:  37552B

Thermo Scientific™  Related applications: Antibody Labeling | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Guardian Peroxidase Conjugate Stabilizer/Diluent preserves the functional integrity and activity of horseradish peroxidase conjugated antibodies and other proteins at very dilute concentrations for long-term storage.

Features of Guardian Peroxidase Conjugate Stabilizer/Diluent:

Preserves HRP activity—no significant loss of HRP activity over a six-month period at room temperature (1:1000 dilution) or 12 months at 4°C
Convenient—store ready-to-use dilutions (1:1000 to 1:100,000) that maintain enzyme activity in the refrigerator—no aliquoting or freezing
Assay compatible—simply add your favorite blocking buffer to create the ideal diluent for your HRP-based ELISA system or store the HRP conjugate as a 1:1000 stock solution for Western blots and dilute in the final assay buffer
Saves money—less expensive than competitor formulations or ordering new HRP conjugates

With Guardian Peroxidase Conjugate Stabilizer/Diluent, typical 1 mg/mL antibody or streptavidin peroxidase conjugates can be diluted as much as 100,000-fold for storage at 4°C. Most ELISA and blotting applications for HRP conjugates require dilution of typical 1 mg/mL stocks by at least 1000-fold (to 1 µg/mL). Sensitive assay systems, such as with chemiluminescent substrates, often require dilution up to 100,000-fold (10 ng/mL). The Guardian Solution enables these 1X working concentrations of HRP conjugate to be prepared in advanced and stored at 4°C for 12 months or at room temperature for six months.For blotting applications, it is usually best to prepare a concentration for storage that can be diluted 10-fold in assay buffer (which may contain blocking components) for final use.

Typical colorimetric ELISA procedures requires that 10 mL of HRP-conjugated secondary antibody be diluted 1000- to 5000-fold from a 1 mg/mL stock solution. That necessitates pipetting 2 to 10 µL of stock into a tube with 10 mL of diluent. More sensitive assay systems, such as Western blots with Thermo Scientific SuperSignal West Pico, Dura or Femto Chemiluminescent Substrates for HRP, require a second round of dilution to achieve the needed working concentration of HRP conjugate. With Guardian Stabilizer-Diluent, HRP conjugates can be pre-diluted and stored at concentrations that are either ready to use or easily diluted 10- to 100-fold for final use. Preparing larger volumes of pre-diluted stocks ensures consistency and greater accuracy for assays performed repeatedly day after day.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: Guardian™
Product Size: Custom
Reagent Type: Diluent, Stabilizing Reagent
Validated Application: ELISA, Western Blot

Contents & storage

Upon receipt store at room temperature or 4°C.

Documents

Manuals & protocols