Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

CAS: 35138-22-8 | C16H24BF4Rh | 406.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3756500101 gGlass bottle
Catalog number 375650010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35138-22-8
IUPAC Nameλ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) tetrafluoroboranuide
Molecular FormulaC16H24BF4Rh
InChI KeyNBGSCPNNKGVTKU-QMDOQEJBSA-N
SMILES[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)Red
Rhodium (Rh)>=24.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only