Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Assorted |
Material | Polycarbonate |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
Unit Size | Pack of 500 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | White |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Case of 2000 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | White |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Pack of 500 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Blue |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Case of 2000 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Blue |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Pack of 500 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Red |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Case of 2000 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Red |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Pack of 500 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Green |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Case of 2000 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Green |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Pack of 500 |
Description | Nunc™ CryoColor Vial Closure Coder, PS |
Color | Yellow |
Material | Polystyrene |
Product Line | Nunc |
Quantity | 500/Pk., 2000/Cs. |
Type | Vial Closure |
For Use With (Equipment) | Cryogenic Vial |
Unit Size | Case of 2000 |
Catalog Number | Specifications | Unit Size | Description | Color | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
375930PK also known as 375930 | Pack of 500 | Nunc™ CryoColor Vial Closure Coder, PS | Assorted | Price: 66.50 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
354755 | Case of 2000 | Nunc™ CryoColor Vial Closure Coder, PS | White | Price: 237.00 Online price: 213.30 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
354755PK also known as 354755 | Pack of 500 | Nunc™ CryoColor Vial Closure Coder, PS | White | Price: 68.70 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
354879 | Case of 2000 | Nunc™ CryoColor Vial Closure Coder, PS | Blue | Price: 241.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
354879PK also known as 354879 | Pack of 500 | Nunc™ CryoColor Vial Closure Coder, PS | Blue | Price: 69.60 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
354968 | Case of 2000 | Nunc™ CryoColor Vial Closure Coder, PS | Red | Price: 241.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
354968PK also known as 354968 | Pack of 500 | Nunc™ CryoColor Vial Closure Coder, PS | Red | Price: 69.60 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
355018 | Case of 2000 | Nunc™ CryoColor Vial Closure Coder, PS | Green | Price: 241.00 Online price: 216.90 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
355018PK also known as 355018 | Pack of 500 | Nunc™ CryoColor Vial Closure Coder, PS | Green | Price: 69.60 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
355077 | Case of 2000 | Nunc™ CryoColor Vial Closure Coder, PS | Yellow | Price: 241.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|