Formic acid hydrazide, 90%
Formic acid hydrazide, 90%
Formic acid hydrazide, 90%
Formic acid hydrazide, 90%
Thermo Scientific Chemicals

Formic acid hydrazide, 90%

CAS: 624-84-0 | CH4N2O | 60.06 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 376061000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS624-84-0
IUPAC Nameformohydrazide
Molecular FormulaCH4N2O
InChI KeyXZBIXDPGRMLSTC-UHFFFAOYSA-N
SMILESNNC=O
さらに表示
Infrared spectrumConforms
Appearance (Color)White to pink
Appearance (Form)Crystalline powder or flakes
Melting point51°C to 59°C
Titration with KIO388 to 112 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only