Ethyl isothiocyanate, 96%
Ethyl isothiocyanate, 96%
Ethyl isothiocyanate, 96%
Thermo Scientific Chemicals

Ethyl isothiocyanate, 96%

CAS: 542-85-8 | C3H5NS | 87.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 376861000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range95% min. (GC)
CAS542-85-8
ChEBICHEBI:85098
Chemical Name or MaterialEthyl isothiocyanate
ColorColorless to Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only